Learn More
Cayman Chemical JWH 302 Analytical Reference S
SDP
Catalog No. 502890369
Encompass_Preferred
A cannabimimetic indole with 5-fold selectivity for the CB1 receptor (Ki = 17 nM) compared to the CB2 receptor (Ki = 89 nM); stimulates GTPγS binding of CB1 and CB2 receptors with EC50 values of 29.3 and 24.4 nM, respectively
Catalog No. 50-289-0369 Supplier Cayman Chemical Supplier No. 1072225 MG
A cannabimimetic indole with 5-fold selectivity for the CB1 receptor (Ki = 17 nM) compared to the CB2 receptor (Ki = 89 nM); stimulates GTPγS binding of CB1 and CB2 receptors with EC50 values of 29.3 and 24.4 nM, respectively