Learn More
Cayman Chemical JWH 307 3-Isomer 10mg
SDP
Catalog No. 502980556
Encompass_Preferred
An isomer of JWH 307 which differs by having the fluorophenyl group at the 3' position, rather than the 2' position, of the pyrrole group; intended for forensic and research purposes
Catalog No. 50-298-0556 Supplier Cayman Chemical Supplier No. 1750610 MG
An isomer of JWH 307 which differs by having the fluorophenyl group at the 3' position, rather than the 2' position, of the pyrrole group; intended for forensic and research purposes