Learn More
Cayman Chemical JWH 307 3-Isomer 1mg
SDP
Catalog No. 502980555
Encompass_Preferred
An isomer of JWH 307 which differs by having the fluorophenyl group at the 3' position, rather than the 2' position, of the pyrrole group; intended for forensic and research purposes
Catalog No. 50-298-0555 Supplier Cayman Chemical Supplier No. 175061 MG
An isomer of JWH 307 which differs by having the fluorophenyl group at the 3' position, rather than the 2' position, of the pyrrole group; intended for forensic and research purposes