Learn More
Cayman Chemical JWH 307 3-Isomer 5mg
SDP
Catalog No. 502980183
Encompass_Preferred
An isomer of JWH 307 which differs by having the fluorophenyl group at the 3' position, rather than the 2' position, of the pyrrole group; intended for forensic and research purposes
Catalog No. 50-298-0183 Supplier Cayman Chemical Supplier No. 175065 MG
An isomer of JWH 307 which differs by having the fluorophenyl group at the 3' position, rather than the 2' position, of the pyrrole group; intended for forensic and research purposes