Learn More
Cayman Chemical JWH 398 N 5hydroxypentyl m
SDP
Catalog No. 502970038
Encompass_Preferred
An internal standard for the quantification of JWH 398 N-(5-hydroxypentyl) metabolite by GC- or LC-MS; intended for forensic and research applications
Catalog No. 50-297-0038 Supplier Cayman Chemical Supplier No. 14371100 UG
An internal standard for the quantification of JWH 398 N-(5-hydroxypentyl) metabolite by GC- or LC-MS; intended for forensic and research applications