Learn More
Cayman Chemical JWH 398 N-pentanoic acid metabolite, Purity: ≥98%, CAS: 1537889-10-3, 10 mg
SDP
Catalog No. 502951379
Encompass_Preferred
An expected phase I metabolite of JWH 398; intended for forensic and research purposes
Catalog No. 50-295-1379 Supplier Cayman Chemical Supplier No. 1094210 MG
An expected phase I metabolite of JWH 398; intended for forensic and research purposes