Learn More
Cayman Chemical JWH398 3CLORNaphthyl Isom 10mg
SDP
Catalog No. 502895512
Encompass_Preferred
Differs structurally from JWH 398 by having chlorine positioned on the naphthyl rings at the 3, rather than the 4, position; intended for forensic purposes
Catalog No. 50-289-5512 Supplier Cayman Chemical Supplier No. 900102410 MG
Differs structurally from JWH 398 by having chlorine positioned on the naphthyl rings at the 3, rather than the 4, position; intended for forensic purposes