Learn More
Cayman Chemical JWH398 7CLORNaphthyl Isom 10mg
SDP
Catalog No. 502895249
Encompass_Preferred
An analytical reference standard that is structurally similar to known synthetic cannabinoids; intended for research and forensic applications
Catalog No. 50-289-5249 Supplier Cayman Chemical Supplier No. 900102710 MG
An analytical reference standard that is structurally similar to known synthetic cannabinoids; intended for research and forensic applications