Learn More
Medchemexpress LLC JYQ-173 | 99.8% | 466.49 | 5 MG
SDP
Catalog No. 5000392575
Encompass_Preferred
JYQ-173 is a chemical probe and a selective inhibitor for human Parkinson disease protein 7 (PARK7), with an IC50 of 19 nM. It exhibits good cell permeability and acts as a ligand for target protein PARK7, which can be utilized for the synthesis of PROTAC Degrader JYQ-194.
  • Selective inhibitor for human Parkinson disease protein 7 (PARK7)
  • Exhibits good cell permeability
  • Can be used as a ligand for the synthesis of PROTAC Degrader
Catalog No. 50-003-92575 Supplier Medchemexpress LLC Supplier No. HY1635635MG
JYQ-173 is a chemical probe and a selective inhibitor for human Parkinson disease protein 7 (PARK7), with an IC50 of 19 nM. It exhibits good cell permeability and acts as a ligand for target protein PARK7, which can be utilized for the synthesis of PROTAC Degrader JYQ-194.
  • Selective inhibitor for human Parkinson disease protein 7 (PARK7)
  • Exhibits good cell permeability
  • Can be used as a ligand for the synthesis of PROTAC Degrader