Learn More
Medchemexpress LLC HY-15249 10mg Medchemexpress, JZL 184 CAS:1101854-58-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1524910MG
Encompass_Preferred
Medchemexpress, HY-15249 10mg JZL 184 CAS:1101854-58-3 JZL 184 is a potent, selective and irreversible monoacylglycerol lipase (MAGL) inhibitor with IC50s of 8 nM and 4 µM for inhibition of MAGL and fatty acid amide hydrolase in mouse brain membranes, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0596