Learn More
Enzo Life Sciences JZL-184 (50mg). CAS: 1101854-58-3
SDP
Supplier:  Enzo Life Sciences BMLEI3910050
Encompass_Preferred
Potent, selective and irreversible inhibitor of MAG lipase (IC50=8nM vs 4µM for FAAH). JZL-184-treated mice displayed a number of CB1-dependent behavioral effects including analgesia, hypothermia and hypomotility as well as increased levels of 2-arachidonoylglycerol in the brain ( approx. 8-fold, with no effect on anandamide levels). JZL-184 produces a rapid and sustained blockade of brain 2-AG hydrolase activity in mice. It blocks neuropathic pain and enhances retrograde endocannabinoid signaling. Alternative name: 4-Nitrophenyl-4-(dibenzo[d][1,3]dioxol-5-yl(hydroxy)methyl)piperidine-1-carboxylate. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (10mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-0768