Learn More
Cayman Chemical JZL 195 InHBTr asy REGE 20nmol
SDP
Catalog No. 502894205
Encompass_Preferred
JZL 195 inhibitor assay reagent has been formulated and tested to work with Cayman’s Assay Kits. Detailed instructions for its use are contained in the respective Cayman Assay Kit booklet.
Catalog No. 50-289-4205 Supplier Cayman Chemical Supplier No. 70030720 NMOL
Add to Cart
Add to Cart
JZL 195 inhibitor assay reagent has been formulated and tested to work with Cayman’s Assay Kits. Detailed instructions for its use are contained in the respective Cayman Assay Kit booklet.