Learn More
Medchemexpress LLC HY-101457 5mg Medchemexpress, JZP-430 CAS:1672691-74-5 Purity:>98%
SDP
Catalog No. 501932138
Encompass_Preferred
Medchemexpress, HY-101457 5mg JZP-430 CAS:1672691-74-5 JZP-430 is a potent, highly selective, irreversible inhibitor of α/β-hydrolase domain 6 (ABHD6) with an IC 50 of 44 nM, exhibits ~230-fold selectivity over fatty acid amide hydrolase (FAAH) and lysosomal acid lipase (LAL) [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2138 Supplier Medchemexpress LLC Supplier No. HY1014575MG
Medchemexpress, HY-101457 5mg JZP-430 CAS:1672691-74-5 JZP-430 is a potent, highly selective, irreversible inhibitor of α/β-hydrolase domain 6 (ABHD6) with an IC 50 of 44 nM, exhibits ~230-fold selectivity over fatty acid amide hydrolase (FAAH) and lysosomal acid lipase (LAL) [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.