Learn More
Cole-Parmer Oakton Buffer Solution, pH 4.01; 1000 mL
Supplier:  Cole-Parmer 0594222
Encompass_Preferred
• Oakton pH Buffer Solutions have pH/temperature tables printed on the labels for accurate calibrations (not on 60-mL, but included as insert) • Manufactured under ISO 9000 quality guidelines and traceable to NIST standards • If unopened, shelf life is 2 years; once opened, use pH 4 and pH 7 within 90 days, pH 10 within 30 days • Buffer Options: - pH 1.68 buffer available in clear - pH 4.01 buffer available in red - pH 6.86 buffer available in clear - pH 7.00 buffer available in yellow - pH 9.18 buffer available in clear - pH 10.01 buffer available in blue - pH 12.46 buffer available in clear
Catalog No. NC1807761