Learn More
Selleck Chemical LLC K-115 hydrochloride dihydrate
SDP
Catalog No. 5000001246
Encompass_Preferred
Ripasudil hydrochloride dihydrate is a potent ROCK inhibitor with IC50 of 51 nM and 19 nM for ROCK1 and ROCK2 respectively used for the treatment of glaucoma and ocular hypertension
Catalog No. 50-000-01246 Supplier Selleck Chemical LLC Supplier No. S799510MM/1ML
Ripasudil hydrochloride dihydrate is a potent ROCK inhibitor with IC50 of 51 nM and 19 nM for ROCK1 and ROCK2 respectively used for the treatment of glaucoma and ocular hypertension