Learn More
Selleck Chemical LLC K-115 hydrochloride dihydrate
Supplier: Selleck Chemical LLC S79951G
Ripasudil hydrochloride dihydrate is a potent ROCK inhibitor with IC50 of 51 nM and 19 nM for ROCK1 and ROCK2 respectively used for the treatment of glaucoma and ocular hypertension
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.