Learn More
AdipoGen K-252a (100 µg)
SDP
Supplier:  AdipoGen AGCN20019C100
Encompass_Preferred
Chemical. CAS: 99533-80-9. Formula: C27H21N3O5. MW: 467.5. Isolated from Nonomuraea longicatena chiba. Antibiotic. PKC, PKA and PKG inhibitor. Shows neurotropic activity by inducing neurite outgrowth. Highly potent cell permeable CaM kinase and phosphorylase inhibitor. Tyrosine protein kinase inhibitor of the TRK family. Antiproliferative. Apoptosis inducer. Cell cycle arrest inducer. Potential drug for psoriasis treatment.
Catalog No. 50-205-3364