Learn More
Medchemexpress LLC HY-N6732 1mg Medchemexpress, K-252a CAS:99533-80-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN67321MG
Encompass_Preferred
Medchemexpress, HY-N6732 1mg K-252a CAS:99533-80-9 K-252a, a staurosporine analog isolated from Nocardiopsis sp. soil fungi, inhibits protein kinase, with IC50 values of 470 nM, 140 nM, 270 nM, and 1.7 nM for PKC, PKA, Ca2+/calmodulin-dependent kinase type II, and phosphorylase kinase, respectively. K-252a is a potent inhibitor (IC50 of 3 nM) of the tyrosine protein kinase (TRK) activity of the NGF receptor gp140trk, the product of the trk protooncogene[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0464