Learn More
Carbosynth LLC. K-252A, 25mg, CAS# 99533-80-9, MF= C27H21N3O5
SDP
Catalog No. 501952408
Encompass_Preferred
Application: Kinase Inhibitor (Pan Specific); Induces Apoptosis and Is Cell Permeable, Notes: K252a is a potent pan-specific protein kinase inhibitor: PKA (Ki = 18 nM), PKC (Ki = 25 nM) PKG (Ki = 20 nM)1. K252a inhibits CaMK (Ki = 1.8 nM) and phosphorylase kinase (IC50 = 1.7 nM) as well as other kinases. Induces apoptosis and is cell permeable.Solubility: Soluble in DMSO (up to 100 mg/ml)., salt: , origin/species: Fermentation product from Nocardiopsis sp., MW: 467.49
Catalog No. 50-195-2408 Supplier Carbosynth LLC. Supplier No. INK3026PI25MG
Application: Kinase Inhibitor (Pan Specific); Induces Apoptosis and Is Cell Permeable, Notes: K252a is a potent pan-specific protein kinase inhibitor: PKA (Ki = 18 nM), PKC (Ki = 25 nM) PKG (Ki = 20 nM)1. K252a inhibits CaMK (Ki = 1.8 nM) and phosphorylase kinase (IC50 = 1.7 nM) as well as other kinases. Induces apoptosis and is cell permeable.Solubility: Soluble in DMSO (up to 100 mg/ml)., salt: , origin/species: Fermentation product from Nocardiopsis sp., MW: 467.49