Learn More
AdipoGen K-252c (5 mg)
SDP
Supplier:  AdipoGen AGCN20097M005
Encompass_Preferred
Chemical. CAS: 85753-43-1. Formula: C20H13N3O. MW: 311.4. Isolated from Streptomyces longisporoflavus. Indolocarbazole alkaloid antibiotic. Potent cell permeable reversible and ATP-competitive PKC (protein kinase C) inhibitor with 10-fold selectivity over PKA (protein kinase A). Anticancer compound. Cytotoxic againt various cancer cell lines. Apoptosis inducer. Antiviral compound against GCV-sensitive and -resistant strains of human cytomegalovirus (HCMV). LACTB (beta-lactamase), malate dehydrogenase (MDH2, MDHC, MDH1B, ME1. ME2, ME3) and chymotrypsin inhibitor. Inhibits mixed lineage kinase (MLK).
Catalog No. 50-205-3454