Learn More
Medchemexpress LLC K-604 dihydrochloride | 217094-32-1 | 99.4% | 575.64 g·mol⁻1 | C23H32Cl2N6OS3 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY100400A10MM 1ML
Encompass_Preferred
K-604 dihydrochloride is a potent, selective acyl-CoA:cholesterol acyltransferase 1 (ACAT-1) inhibitor used in biochemical and cellular research to study cholesterol esterification and related pathways. It is supplied as a 10 mM solution in DMSO or as a solid for preparation of assay stocks and controls.
  • Potent, selective ACAT-1 inhibition with low micromolar IC50.
  • Suitable for biochemical and cellular assays.
  • Available as a 10 mM solution in DMSO and in solid pack sizes.
  • High purity for research applications.
  • Includes CAS, molecular formula, and molecular weight for identification.
Catalog No. 50-002-13922