Learn More
TARGETMOL CHEMICALS INC K-604 dihydrochloride 5MG
SDP
Catalog No. 502880697
Encompass_Preferred
Also available in 1 mL, 1 mg, 2 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. K-604 dihydrochloride, a potent and selective acyl-CoAcholesterol acyltransferase 1 inhibitor, exhibits an IC50 of 0.45±0.06 uM. Purity 99.88%
Catalog No. 50-288-0697 Supplier TARGETMOL CHEMICALS INC Supplier No. T117335MG
Also available in 1 mL, 1 mg, 2 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. K-604 dihydrochloride, a potent and selective acyl-CoAcholesterol acyltransferase 1 inhibitor, exhibits an IC50 of 0.45±0.06 uM. Purity 99.88%