Learn More
Selleck Chemical LLC K-604 dihydrochloride-5MG
SDP
Catalog No. 503135887
Encompass_Preferred
K-604 dihydrochloride a potent and selective inhibitor of Acyl-coenzyme A cholesterol O-acyltransferase 1 (ACAT1) suppresses the development of atherosclerosis in an animal model without affecting plasma cholesterol levels
Catalog No. 50-313-5887 Supplier Selleck Chemical LLC Supplier No. E09415MG
K-604 dihydrochloride a potent and selective inhibitor of Acyl-coenzyme A cholesterol O-acyltransferase 1 (ACAT1) suppresses the development of atherosclerosis in an animal model without affecting plasma cholesterol levels