Learn More
Cayman Chemical K-7174 (hydrochloride), Purity: ≥98%, CAS: 191089-60-8, 5 mg
SDP
Catalog No. 502950011
Encompass_Preferred
A 20S proteasome inhibitor; inhibits the 20S proteasome β1, β2, and β5 catalytic subunits at 5 µM; reduces TNF-α- or IL-1β-induced adherence of U937 monocytes to HUVECs at 10 µM; inhibits tumor growth in U266 and RPMI-8226 multiple myeloma mouse xenograft models at 75 mg/kg; an inhibitor of the GATA consensus sequence
Catalog No. 50-295-0011 Supplier Cayman Chemical Supplier No. 327735 MG
A 20S proteasome inhibitor; inhibits the 20S proteasome β1, β2, and β5 catalytic subunits at 5 µM; reduces TNF-α- or IL-1β-induced adherence of U937 monocytes to HUVECs at 10 µM; inhibits tumor growth in U266 and RPMI-8226 multiple myeloma mouse xenograft models at 75 mg/kg; an inhibitor of the GATA consensus sequence