Learn More
Medchemexpress LLC K-80003 | 1292821-90-9 | 336.40 | 50 MG
SDP
Catalog No. 5000446841
Encompass_Preferred
K-80003 is a potent inhibitor of tRXRα-dependent Akt activation and cancer cell growth. It demonstrates high affinity to RXRα and lacks COX inhibitory activity, displaying enhanced efficacy in inhibiting tRXRα-dependent AKT activation and tumor growth in animals.
  • Inhibits TNFα-induced colocalization of tRXRα with p85α
  • Stabilizes tetrameric tRXRα in the nuclear fraction
  • Potent inhibitor of AKT activation
  • Enhanced efficacy in inhibiting tRXRα-dependent AKT activation and tumor growth in animals
  • High affinity to RXRα
  • Lacks COX inhibitory activity
Catalog No. 50-004-46841 Supplier Medchemexpress LLC Supplier No. HYU0045850MG
K-80003 is a potent inhibitor of tRXRα-dependent Akt activation and cancer cell growth. It demonstrates high affinity to RXRα and lacks COX inhibitory activity, displaying enhanced efficacy in inhibiting tRXRα-dependent AKT activation and tumor growth in animals.
  • Inhibits TNFα-induced colocalization of tRXRα with p85α
  • Stabilizes tetrameric tRXRα in the nuclear fraction
  • Potent inhibitor of AKT activation
  • Enhanced efficacy in inhibiting tRXRα-dependent AKT activation and tumor growth in animals
  • High affinity to RXRα
  • Lacks COX inhibitory activity