Learn More
TARGETMOL CHEMICALS INC K-80003 5MG
SDP
Catalog No. 502581514
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. K-80003(TX-803) is a potent retinol-like X-receptor-alpha regulator that inhibits TRxr(alpha)-dependent Akt activation and tumor cell growth. purity: 99%
Catalog No. 50-258-1514 Supplier TARGETMOL CHEMICALS INC Supplier No. T117345MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. K-80003(TX-803) is a potent retinol-like X-receptor-alpha regulator that inhibits TRxr(alpha)-dependent Akt activation and tumor cell growth. purity: 99%