Learn More
Med Vet International K-9 EZ Boy Radiographic Positioning Aid, Non-Covered, Small, J1287, Each
Catalog No. 5000581088
Encompass_Preferred
K-9 EZ Boy. Radiographic Positioning Aids. Positioning aids are valuable in: - Reducing radiation exposure to clinic staff. - Decreasing motion. - Positioning animals for a variety of procedures. - Patient comfort. Radiographic Positioning Aids are available in a variety of shapes with the most popular being the 'V' trough. Two Styles 1. Non-coated: soft open cell gray sponge material provides excellent support and comfort at a less expensive cost. 2. Covered: dark blue nylon reinforced vinyl. Also features hook and loop straps to secure limbs; readily cleaned for quick re-use. Non-Covered. Small. Sizes: Length / Width / Height Small 10" 4" 3" Medium 15" 7" 5" Large 24" 11" 7" X-Large 36" 14" 9" Actual Weight:  .15 lbs. Weight reflected in additional information is in reference to dimensional weight for shipping purposes.
Catalog No. 50-005-81088 Supplier Med Vet International Supplier No. JOR1287
K-9 EZ Boy. Radiographic Positioning Aids. Positioning aids are valuable in: - Reducing radiation exposure to clinic staff. - Decreasing motion. - Positioning animals for a variety of procedures. - Patient comfort. Radiographic Positioning Aids are available in a variety of shapes with the most popular being the 'V' trough. Two Styles 1. Non-coated: soft open cell gray sponge material provides excellent support and comfort at a less expensive cost. 2. Covered: dark blue nylon reinforced vinyl. Also features hook and loop straps to secure limbs; readily cleaned for quick re-use. Non-Covered. Small. Sizes: Length / Width / Height Small 10" 4" 3" Medium 15" 7" 5" Large 24" 11" 7" X-Large 36" 14" 9" Actual Weight:  .15 lbs. Weight reflected in additional information is in reference to dimensional weight for shipping purposes.