Learn More
Jorgensen Labs K-9 STOMACH TUBE MEDIUM
SDP
Catalog No. NC0456766
Encompass_Preferred
Larger size PVC stomach tube for bigger dogs. Useful in gastric lavage or administering larger volumes of fluids with special funnel adaptor. Plus has one lateral eye. Medium. 1/ 4 in. i.d. x 3/ 8 in. o.d. x 31/2c. For dogs > 40 lbs
Catalog No. NC0456766 Supplier Jorgensen Labs Supplier No. J0348Q
Larger size PVC stomach tube for bigger dogs. Useful in gastric lavage or administering larger volumes of fluids with special funnel adaptor. Plus has one lateral eye. Medium. 1/ 4 in. i.d. x 3/ 8 in. o.d. x 31/2c. For dogs > 40 lbs