Learn More
Selleck Chemical LLC K-975-E1329-5MG
SDP
Catalog No. 502389924
Encompass_Preferred
K-975 is a potent selective and orally active TEAD inhibitor that directly inhibits YAP/TAZ-TEAD protein-protein interaction and shows a potent anti-tumor effect in malignant pleural mesothelioma
Catalog No. 50-238-9924 Supplier Selleck Chemical LLC Supplier No. E13295MG
K-975 is a potent selective and orally active TEAD inhibitor that directly inhibits YAP/TAZ-TEAD protein-protein interaction and shows a potent anti-tumor effect in malignant pleural mesothelioma