Learn More
Sino Biological K Cadherin / CDH6 Antibody, Mouse MAb 100uL
Supplier:  Sino Biological 10150MM09100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human K Cadherin / CDH6 (rh K Cadherin / CDH6; Catalog#10150-H08H; NP_004923.1; Met1-Ala615). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-224-8502