Learn More
YSI 3P LifeScience K FERROCYANIDE
Catalog No. 50200151
Encompass
Potassium Ferrocyanide (125 mL). Use to check membrane integrity on YSI 1500, 2300, 2700, 2900 Series and 7100 analyzers. 14459-95-1
Catalog No. 50-200-151 Supplier YSI 3P LifeScience Supplier No. 087052
Potassium Ferrocyanide (125 mL). Use to check membrane integrity on YSI 1500, 2300, 2700, 2900 Series and 7100 analyzers. 14459-95-1