Learn More
New England Biolabs, Inc. K. lactis GG799 Competent Cells – 5 transformation reactions
SDP
Catalog No. 501526335
Encompass
Chemically competent Kluyveromyces lactis cells suitable for transformation with a linearized pKLAC1-based expression vector. GG799 cells are characterized by very high cell density growth and efficient expression of foreign proteins.
Catalog No. 50-152-6335 Supplier New England Biolabs, Inc. Supplier No. C1001S
Chemically competent Kluyveromyces lactis cells suitable for transformation with a linearized pKLAC1-based expression vector. GG799 cells are characterized by very high cell density growth and efficient expression of foreign proteins.