Learn More
New England Biolabs, Inc. K.lactis Protein Expression Kit – 1 set
SDP
Catalog No. 50994879
Encompass
The K. lactis Protein Expression Kit provides an easy method for expressing a gene of interest in the yeast Kluyveromyces lactis. Proteins may be produced intracellularly or secreted using an integrative expression vector (pKLAC2). To achieve protein secretion, a gene of interest is cloned downstream of the K. lactis -mating factor secretion domain, which is eventually processed in the Golgi and results in secretion of the desired protein.
Catalog No. 50-994-879 Supplier New England Biolabs, Inc. Supplier No. E1000S
The K. lactis Protein Expression Kit provides an easy method for expressing a gene of interest in the yeast Kluyveromyces lactis. Proteins may be produced intracellularly or secreted using an integrative expression vector (pKLAC2). To achieve protein secretion, a gene of interest is cloned downstream of the K. lactis -mating factor secretion domain, which is eventually processed in the Golgi and results in secretion of the desired protein.