Learn More
Medchemexpress LLC HY-N0628 1mg Medchemexpress, Kaempferitrin CAS:482-38-2 Purity:>98%
SDP
Catalog No. 501933647
Encompass_Preferred
Medchemexpress, HY-N0628 1mg Kaempferitrin CAS:482-38-2 Kaempferitrin is a natural flavonoid, possesses antinociceptive, anti-inflammatory, anti-diabetic, antitumoral and chemopreventive effects, and activates insulin signaling pathway. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3647 Supplier Medchemexpress LLC Supplier No. HYN06281MG
Add to Cart
Add to Cart
Medchemexpress, HY-N0628 1mg Kaempferitrin CAS:482-38-2 Kaempferitrin is a natural flavonoid, possesses antinociceptive, anti-inflammatory, anti-diabetic, antitumoral and chemopreventive effects, and activates insulin signaling pathway. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.