Learn More
Enzo Life Sciences Kaempferol (50mg). CAS: 520-18-3
SDP
Supplier:  Enzo Life Sciences ALX385005M050
Encompass_Preferred
Antioxidant flavonoid. Apoptosis inducer. Reversible inhibitor of fatty acid synthase (FAS). Alternative name: 3,4',5,7-Tetrahydroxyflavone. Purity: ≥90% (HPLC). Appearance: Yellow powder. Solubility: Soluble in ether or 100% ethanol (hot); slightly soluble in water. Long Term Storage: +4°C.
Catalog No. 50-200-9038