Learn More
J Kem Scientific Inc K PROBE 1/8X18IN
SDP
Catalog No. NC1592457
Encompass
Single-Element Thermocouple | Type K | Teflon Overcoating | Diameter: 1/8" | Length 18"
Catalog No. NC1592457 Supplier J Kem Scientific Inc Supplier No. K1/8T18
Single-Element Thermocouple | Type K | Teflon Overcoating | Diameter: 1/8" | Length 18"