Learn More
Selleck Chemical LLC K-Ras
SDP
Supplier:  Selleck Chemical LLC S733310MG
Encompass_Preferred
K-Ras(G12C) inhibitor 6 is an allosteric and selective inhibitor of oncogenic K-Ras(G12C)
Catalog No. 50-000-04587