Learn More
Medchemexpress LLC K-Ras G12C-IN-4 | 2376328-55-9 | 99.2% | 50 MG
SDP
Catalog No. 5000340855
Encompass_Preferred
K-Ras G12C-IN-4 is a potent covalent inhibitor of KRASG12C. It is intended for research use only. This compound exhibits significant cytotoxic activity against human A549 cells and MIAPaCa2 cells, with IC50 values of 9.33 μM and 0.067 μM, respectively. It also inhibits MAPK signaling (p-ERK) in MIA PaCa-2 cells with an IC50 of 0.219 μM.
  • Potent covalent inhibitor of KRASG12C
  • For research use only
  • Inhibits cytotoxicity against human A549 cells (IC50: 9.33 μM)
  • Inhibits cytotoxicity against human MIAPaCa2 cells (IC50: 0.067 μM)
  • Inhibits MAPK signaling (p-ERK) in MIA PaCa-2 cells (IC50: 0.219 μM)
Catalog No. 50-003-40855 Supplier Medchemexpress LLC Supplier No. HY12877150MG
Add to Cart
Add to Cart
K-Ras G12C-IN-4 is a potent covalent inhibitor of KRASG12C. It is intended for research use only. This compound exhibits significant cytotoxic activity against human A549 cells and MIAPaCa2 cells, with IC50 values of 9.33 μM and 0.067 μM, respectively. It also inhibits MAPK signaling (p-ERK) in MIA PaCa-2 cells with an IC50 of 0.219 μM.
  • Potent covalent inhibitor of KRASG12C
  • For research use only
  • Inhibits cytotoxicity against human A549 cells (IC50: 9.33 μM)
  • Inhibits cytotoxicity against human MIAPaCa2 cells (IC50: 0.067 μM)
  • Inhibits MAPK signaling (p-ERK) in MIA PaCa-2 cells (IC50: 0.219 μM)