Learn More
TARGETMOL CHEMICALS INC K-RAS G12C-IN-4 5MG
SDP
Catalog No. 502577446
Encompass_Preferred
Also available in 1mg 2mg 10mg 25mg 50mg 100mg and bulk. Please contact Fisher for quotes. K-Ras G12C-IN-4 is a potent co-valent Inhibitor of KRAS G12C.. purity: 99%
Catalog No. 50-257-7446 Supplier TARGETMOL CHEMICALS INC Supplier No. T117385MG
Add to Cart
Add to Cart
Also available in 1mg 2mg 10mg 25mg 50mg 100mg and bulk. Please contact Fisher for quotes. K-Ras G12C-IN-4 is a potent co-valent Inhibitor of KRAS G12C.. purity: 99%