Learn More
Apexbio Technology LLC K-Ras(G12C) inhibitor 12 1469337-95-8 5mg
SDP
Catalog No. 5000087943
Encompass_Preferred
K-Ras(G12C) inhibitor 12 (CAS 1469337-95-8) is an allosteric inhibitor targeting the K-Ras(G12C) mutant a GTPase implicated in various signal transduction pathways and commonly mutated in cancers This small molecule reduces the affinity of K-Ras(G12C) for GTP promotes covalent modification of the mutant protein and inhibits SOS-mediated nucleotide exchange In K-Ras(G12C)-mutant lung cancer cell lines (H1792 H358 H23 Calu-1) it disrupts C-Raf and B-Raf association with Ras decreases cell viability and induces apoptosis In H1792 cells the EC50 is 0 32 M K-Ras(G12C) inhibitor 12 is valuable for studies of Ras biology and targeted cancer research
Catalog No. 50-000-87943 Supplier Apexbio Technology LLC Supplier No. B48765
Add to Cart
Add to Cart
K-Ras(G12C) inhibitor 12 (CAS 1469337-95-8) is an allosteric inhibitor targeting the K-Ras(G12C) mutant a GTPase implicated in various signal transduction pathways and commonly mutated in cancers This small molecule reduces the affinity of K-Ras(G12C) for GTP promotes covalent modification of the mutant protein and inhibits SOS-mediated nucleotide exchange In K-Ras(G12C)-mutant lung cancer cell lines (H1792 H358 H23 Calu-1) it disrupts C-Raf and B-Raf association with Ras decreases cell viability and induces apoptosis In H1792 cells the EC50 is 0 32 M K-Ras(G12C) inhibitor 12 is valuable for studies of Ras biology and targeted cancer research