Learn More
Medchemexpress LLC K-Ras(G12C) inhibitor 12 | 1469337-95-8 | 99.6% | 449.67 | 100 MG
SDP
Catalog No. 5000388057
Encompass_Preferred
K-Ras(G12C) inhibitor 12 is an irreversible inhibitor of K-Ras(G12C) that can alter the nucleotide-binding preference of K-Ras and block its interaction with effector proteins. It can reduce cell viability and induce apoptosis in lung cancer cell lines with G12C mutations and has anti-tumor activity.
  • Irreversible inhibitor of K-Ras(G12C)
  • Alters nucleotide-binding preference of K-Ras
  • Blocks interaction with effector proteins
  • Reduces cell viability and induces apoptosis in lung cancer cell lines with G12C mutations
  • Has anti-tumor activity
Catalog No. 50-003-88057 Supplier Medchemexpress LLC Supplier No. HY18707100MG
K-Ras(G12C) inhibitor 12 is an irreversible inhibitor of K-Ras(G12C) that can alter the nucleotide-binding preference of K-Ras and block its interaction with effector proteins. It can reduce cell viability and induce apoptosis in lung cancer cell lines with G12C mutations and has anti-tumor activity.
  • Irreversible inhibitor of K-Ras(G12C)
  • Alters nucleotide-binding preference of K-Ras
  • Blocks interaction with effector proteins
  • Reduces cell viability and induces apoptosis in lung cancer cell lines with G12C mutations
  • Has anti-tumor activity