Learn More
Medchemexpress LLC K-Ras(G12C) inhibitor 12 | 1469337-95-8 | 99.6% | 449.67 | 5 MG
SDP
Catalog No. 5000386111
Encompass_Preferred
K-Ras(G12C) inhibitor 12 is an irreversible inhibitor of K-Ras(G12C). It can alter the nucleotide-binding preference of K-Ras and block its interaction with effector proteins. K-Ras(G12C) inhibitor 12 can reduce cell viability and induce apoptosis in lung cancer cell lines with G12C mutations and has anti-tumor activity. It is for research use only.
  • Irreversible inhibitor of K-Ras(G12C)
  • Reduces cell viability and induces apoptosis in lung cancer cell lines with G12C mutations
  • Has anti-tumor activity
  • Appears as a solid, white to light yellow
Catalog No. 50-003-86111 Supplier Medchemexpress LLC Supplier No. HY187075MG
Add to Cart
Add to Cart
K-Ras(G12C) inhibitor 12 is an irreversible inhibitor of K-Ras(G12C). It can alter the nucleotide-binding preference of K-Ras and block its interaction with effector proteins. K-Ras(G12C) inhibitor 12 can reduce cell viability and induce apoptosis in lung cancer cell lines with G12C mutations and has anti-tumor activity. It is for research use only.
  • Irreversible inhibitor of K-Ras(G12C)
  • Reduces cell viability and induces apoptosis in lung cancer cell lines with G12C mutations
  • Has anti-tumor activity
  • Appears as a solid, white to light yellow