Learn More
Apexbio Technology LLC K-Ras(G12C) inhibitor 9 1469337-91-4 5mg
SDP
Catalog No. 5000092406
Encompass_Preferred
K-Ras(G12C) inhibitor 9 (CAS 1469337-91-4) is a covalent small molecule that selectively targets the oncogenic K-Ras(G12C) mutant protein Acting as an allosteric inhibitor it irreversibly binds to the mutant K-Ras interfering with the binding of GTP and GDP and disrupts interactions with effector proteins such as B-Raf and C-Raf In vitro studies show that treatment with 10 M K-Ras(G12C) inhibitor 9 reduces viability and increases apoptosis in K-Ras(G12C)-mutant lung cancer cell lines including H1792 Calu-1 H358 and H23 This compound is a valuable research tool for investigating K-Ras-driven cancer signaling and mutant-selective therapeutic strategies
Catalog No. 50-000-92406 Supplier Apexbio Technology LLC Supplier No. B60025
Add to Cart
Add to Cart
K-Ras(G12C) inhibitor 9 (CAS 1469337-91-4) is a covalent small molecule that selectively targets the oncogenic K-Ras(G12C) mutant protein Acting as an allosteric inhibitor it irreversibly binds to the mutant K-Ras interfering with the binding of GTP and GDP and disrupts interactions with effector proteins such as B-Raf and C-Raf In vitro studies show that treatment with 10 M K-Ras(G12C) inhibitor 9 reduces viability and increases apoptosis in K-Ras(G12C)-mutant lung cancer cell lines including H1792 Calu-1 H358 and H23 This compound is a valuable research tool for investigating K-Ras-driven cancer signaling and mutant-selective therapeutic strategies