Learn More
Cayman Chemical K-Ras(G12C) Inhibitor 12, Purity: ≥95%, CAS: 1469337-95-8, 25 mg
SDP
Catalog No. 502982762
Encompass_Preferred
An allosteric inhibitor of oncogenic K-Ras(G12C); decreases viability and increases apoptosis in several lung cancer cell lines that contain G12C mutations (EC50 = 0.32 µM in H1792 cells)
Catalog No. 50-298-2762 Supplier Cayman Chemical Supplier No. 2011825 MG
An allosteric inhibitor of oncogenic K-Ras(G12C); decreases viability and increases apoptosis in several lung cancer cell lines that contain G12C mutations (EC50 = 0.32 µM in H1792 cells)