Learn More
Cayman Chemical K-Ras(G12C) Inhibitor 9, Purity: ≥98%, CAS: 1469337-91-4, 10 mg
SDP
Catalog No. 502966822
Encompass_Preferred
An irreversible, allosteric inhibitor of the K-Ras(G12C) mutant that causes 100% modification of the protein when used at 10 µM for 24 hours in vitro
Catalog No. 50-296-6822 Supplier Cayman Chemical Supplier No. 1800510 MG
An irreversible, allosteric inhibitor of the K-Ras(G12C) mutant that causes 100% modification of the protein when used at 10 µM for 24 hours in vitro