Learn More
Cayman Chemical K-Ras Isoform AG12C mutnt100ug
SDP
Catalog No. 502942963
Encompass_Preferred
Source: Recombinant human N-terminal His-tagged K-RasG12C isoform A expressed in E. coli • Amino acids: 2-186 • MW: 23 kDa
Catalog No. 50-294-2963 Supplier Cayman Chemical Supplier No. 40371100 UG
Source: Recombinant human N-terminal His-tagged K-RasG12C isoform A expressed in E. coli • Amino acids: 2-186 • MW: 23 kDa