Learn More
Cayman Chemical K-Ras Isoform AG12D mutnt100ug
SDP
Catalog No. 502943249
Encompass_Preferred
Recombinant human N-terminal His-tagged K-Ras(G12D) isoform A expressed in E. coli • Amino acids: 2-186 • MW: 23 kDa
Catalog No. 50-294-3249 Supplier Cayman Chemical Supplier No. 40372100 UG
Recombinant human N-terminal His-tagged K-Ras(G12D) isoform A expressed in E. coli • Amino acids: 2-186 • MW: 23 kDa