Learn More
Cayman Chemical K-Ras Isoform BG12CC51S100ug
SDP
Catalog No. 502942323
Encompass_Preferred
Source: Recombinant human N-Terminal His-tagged K-Ras Isoform B (G12C, C51S, C80L, C118S) expressed in E. coli • Amino acids: 1-169 • MW: 22.27 kDa
Catalog No. 50-294-2323 Supplier Cayman Chemical Supplier No. 40366100 UG
Source: Recombinant human N-Terminal His-tagged K-Ras Isoform B (G12C, C51S, C80L, C118S) expressed in E. coli • Amino acids: 1-169 • MW: 22.27 kDa