Learn More
Cayman Chemical K-Ras Isoform BG12V mutnt100ug
SDP
Catalog No. 502945384
Encompass_Preferred
Source: Active recombinant human N-terminal His-tagged K-Ras(G12V) isoform B expressed in insect cells • Amino acids: 2-185 • MW: 22 kDa
Catalog No. 50-294-5384 Supplier Cayman Chemical Supplier No. 40373100 UG
Source: Active recombinant human N-terminal His-tagged K-Ras(G12V) isoform B expressed in insect cells • Amino acids: 2-185 • MW: 22 kDa