Learn More
Medchemexpress LLC HY-130707 1mg Medchemexpress, K-Ras ligand-Linker Conjugate 3 CAS:2378261-87-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1307071MG
Encompass_Preferred
Medchemexpress, HY-130707 1mg K-Ras ligand-Linker Conjugate 3 CAS:2378261-87-9 K-Ras ligand-Linker Conjugate 3 (Compound 001371) incorporates a ligand for FAK (a KRas recruiting moiety), and a PROTAC linker, which recruit E3 ligases (such as VHL, CRBN, MDM2, and IAP). K-Ras ligand-Linker Conjugate 3 can be extensively used for PROTAC-mediated protein degradation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9833